Thermo Scientific Chemicals

trans-4-(4-n-Pentylcyclohexyl)benzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: H57351.03
1 g, Each
Thermo Scientific Chemicals

trans-4-(4-n-Pentylcyclohexyl)benzoic acid, 99%, Thermo Scientific Chemicals

Catalog number: H57351.03
1 g, Each
Quantity
Catalog number: H57351.03
also known as H57351-03
Price (USD)
Price: 38.70
Online price: 33.65
Your price:
Quantity
-

Chemical Identifiers

CAS
65355-30-8
IUPAC Name
4-(4-pentylcyclohexyl)benzoic acid
Molecular Formula
C18H26O2
InChI Key
YXKKMVGGPRVHIL-UHFFFAOYSA-N
SMILES
CCCCCC1CCC(CC1)C1=CC=C(C=C1)C(O)=O
Assay (unspecified)
>98.5%

Description

It has been used for intermediate of liquid crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been used for intermediate of liquid crystals.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text