Thermo Scientific Chemicals

N-Boc-trans-4-hydroxy-L-proline methyl ester, 97%, Thermo Scientific Chemicals

Catalog number: H59027.03
1 g, Each
Thermo Scientific Chemicals

N-Boc-trans-4-hydroxy-L-proline methyl ester, 97%, Thermo Scientific Chemicals

Catalog number: H59027.03
1 g, Each
Quantity
Catalog number: H59027.03
also known as H59027-03
Price (USD)
Price: 35.70
Online price: 30.65
Your price:
Quantity
-

Chemical Identifiers

CAS
74844-91-0
IUPAC Name
1-tert-butyl 2-methyl (2S,4R)-4-hydroxypyrrolidine-1,2-dicarboxylate
Molecular Formula
C11H19NO5
InChI Key
MZMNEDXVUJLQAF-SFYZADRCSA-N
SMILES
COC(=O)[C@@H]1C[C@@H](O)CN1C(=O)OC(C)(C)C
Appearance (Color)
White to yellow
Form
Powder
Proton NMR
>96.0%

Description

It is used in the synthesis of For-Met-Leu-Phe-OMe (fMLF-OMe) analogues based on Met residue replacement by 4-amino-proline scaffold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of For-Met-Leu-Phe-OMe (fMLF-OMe) analogues based on Met residue replacement by 4-amino-proline scaffold.

Solubility
Soluble in chloroform, dichloromethane and ethyl acetate.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text