Thermo Scientific Chemicals

4-Hydroxybenzenesulfonic acid, 65%, Thermo Scientific Chemicals

Catalog number: H59917.K2
1 L, Each
Thermo Scientific Chemicals

4-Hydroxybenzenesulfonic acid, 65%, Thermo Scientific Chemicals

Catalog number: H59917.K2
1 L, Each
Quantity
Catalog number: H59917.K2
also known as H59917-K2
Price (USD)
Price: 190.00
Online price: 161.65
Your price:
Quantity
-

Chemical Identifiers

CAS
98-67-9
Concentration (weight/weight)
65 wt. % in H2O

Description

4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxybenzenesulfonic Acid is used as a redox-mediator in the laccase-catalyzed degradation of indigo, a textile dye. Also used in oilfield chemicals, metal working, and as a polymerization catalyst.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text