Thermo Scientific Chemicals

Methyl piperidine-4-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H61754.14
25 g, Each
Thermo Scientific Chemicals

Methyl piperidine-4-carboxylate, 98%, Thermo Scientific Chemicals

Catalog number: H61754.14
25 g, Each
Quantity
Catalog number: H61754.14
also known as H61754-14
Price (USD)
Price: 53.20
Online price: 45.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2971-79-1
IUPAC Name
methyl piperidine-4-carboxylate
Molecular Formula
C7H13NO2
InChI Key
RZVWBASHHLFBJF-UHFFFAOYSA-N
SMILES
COC(=O)C1CCNCC1
Assay (GC)
>97.5%

Description

Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl piperidine-4-carboxylate is a reactant for C-2 arylation of piperidines through directed transition metal-catalyzed sp3 C-H activation. It is used as a reactant for synthesis of antitubercular agents, aminopyrazine inhibitors, orally available naphthyridine protein kinase D inhibitors and a scalable catalyst from asymmetric hydrogenation of sterically demanding aryl enamide.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text