Thermo Scientific Chemicals

3-Fluoropyridine-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H61902.03
1 g, Each
Thermo Scientific Chemicals

3-Fluoropyridine-2-carboxaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H61902.03
1 g, Each
Quantity
Catalog number: H61902.03
also known as H61902-03
Price (USD)
Price: 92.70
Your price:
Quantity
-

Chemical Identifiers

CAS
31224-43-8
IUPAC Name
3-fluoropyridine-2-carbaldehyde
Molecular Formula
C6H4FNO
InChI Key
OZIMPUNGBUYCSP-UHFFFAOYSA-N
SMILES
FC1=C(C=O)N=CC=C1
Assay (GC)
>97.5%

Description

3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoropyridine-2-carboxaldehyde is used to prepare taxoids derived from 9β-dihydrobaccatin-9,10-acetals with anti-tumor activities. It is also an intermediate used to synthesize S-3578 derivatives with potent activities against both methicillin-resistant Staphylococcus aureus (MRSA) and Pseudomonas aeruginosa.

Solubility
Slightly soluble in water.

Notes
Store in cool, well ventilated area. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text