Thermo Scientific Chemicals

3-Hydroxypyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H63465.03
1 g, Each
Thermo Scientific Chemicals

3-Hydroxypyridine-4-carboxylic acid, 98%, Thermo Scientific Chemicals

Catalog number: H63465.03
1 g, Each
Quantity
Catalog number: H63465.03
also known as H63465-03
Price (USD)
Price: 29.60
Online price: 25.65
Your price:
Quantity
-

Chemical Identifiers

CAS
10128-71-9
IUPAC Name
3-hydroxypyridine-4-carboxylate
Molecular Formula
C6H4NO3
InChI Key
JEHGATQUCUYHJL-UHFFFAOYSA-M
SMILES
OC1=CN=CC=C1C([O-])=O
Proton NMR
>97.5%

Description

3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Hydroxyisonicotinic Acid is used in the preparation of chelating agents for iron and aluminum.

Solubility
Soluble in methanol. Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text