Thermo Scientific Chemicals

3-Bromo-4-methoxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: H64296.06
5 g, Each
Thermo Scientific Chemicals

3-Bromo-4-methoxypyridine, 97%, Thermo Scientific Chemicals

Catalog number: H64296.06
5 g, Each
Quantity
Catalog number: H64296.06
also known as H64296-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
82257-09-8
IUPAC Name
3-bromo-4-methoxypyridine
Molecular Formula
C6H6BrNO
InChI Key
DSFJKFHIMBVWAV-UHFFFAOYSA-N
SMILES
COC1=C(Br)C=NC=C1
Assay (HPLC)
>96.0%

Description

3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-4-methoxypyridine acid is used as a intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water (9.8 g/L).

Notes
Store in freezer at (-20°C). Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text