Thermo Scientific Chemicals

4-Bromo-3-chloro-2-fluoropyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64362.03
1 g, Each
Thermo Scientific Chemicals

4-Bromo-3-chloro-2-fluoropyridine, 98%, Thermo Scientific Chemicals

Catalog number: H64362.03
1 g, Each
Quantity
Catalog number: H64362.03
also known as H64362-03
Price (USD)
Quantity
-

Chemical Identifiers

CAS
1017793-21-3
IUPAC Name
4-bromo-3-chloro-2-fluoropyridine
Molecular Formula
C5H2BrClFN
InChI Key
UMQBGWKWOLRROL-UHFFFAOYSA-N
SMILES
FC1=NC=CC(Br)=C1Cl
Assay (HPLC)
>97.5%

Description

Useful intermediate for pharmaceutical and organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text