Thermo Scientific Chemicals

4-Iodo-o-phenylenediamine, 95%, Thermo Scientific Chemicals

Catalog number: H64469.03
1 g, Each
Thermo Scientific Chemicals

4-Iodo-o-phenylenediamine, 95%, Thermo Scientific Chemicals

Catalog number: H64469.03
1 g, Each
Quantity
Catalog number: H64469.03
also known as H64469-03
Price (USD)
Price: 123.00
Online price: 106.65
Your price:
Quantity
-

Chemical Identifiers

CAS
21304-38-1
IUPAC Name
4-iodobenzene-1,2-diamine
Molecular Formula
C6H7IN2
InChI Key
FUOSRKZBOIVBOS-UHFFFAOYSA-N
SMILES
NC1=C(N)C=C(I)C=C1
Assay (HPLC)
>94.0%

Description

It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis of N-(Benzimidazol-2-ylmethyl)iminodiacetic acids.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from light, acids and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text