Benzenesulfonic acid isopropyl ester, 95%
Benzenesulfonic acid isopropyl ester, 95%
Benzenesulfonic acid isopropyl ester, 95%
Thermo Scientific Chemicals

Benzenesulfonic acid isopropyl ester, 95%

CAS: 6214-18-2 | C9H12O3S | 200.252 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number H64523.03
also known as H64523-03
Price (USD)
49.40
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
49.40
Each
Chemical Identifiers
CAS6214-18-2
IUPAC Name2-methylpropyl benzenesulfonate
Molecular FormulaC10H14O3S
InChI KeyNFRRFEFZKGNYKS-UHFFFAOYSA-N
SMILESCC(C)COS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay (unspecified)≥94.0%
A sulfonate ester that acts as a potential effects in drug substances impurity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A sulfonate ester that acts as a potential effects in drug substances impurity.

Solubility
Soluble in chloroform and ethyl acetate.

Notes
Store at 4°C. Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
RUO – Research Use Only

General References:

  1. Guo T.; Shi, Y.; Zheng, L., et al. Rapid and simultaneous determination of sulfonate ester genotoxic impurities in drug substance by liquid chromatography coupled to tandem mass spectrometry: Comparison of different ionization modes. J Chromatogr A. 2014 135573-79.
  2. John N. Bonfigliol. Directed ortho-lithiation of alkyl arenesulfonates. J. Org. Chem. 1986 51(14), 2833-2835.