Thermo Scientific Chemicals

2-Bromo-6-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H64689.06
5 g, Each
Thermo Scientific Chemicals

2-Bromo-6-fluorobenzaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: H64689.06
5 g, Each
Quantity
Catalog number: H64689.06
also known as H64689-06
Price (USD)
Price: 39.70
Online price: 33.65
Your price:
Quantity
-

Chemical Identifiers

CAS
360575-28-6
IUPAC Name
2-bromo-6-fluorobenzaldehyde
Molecular Formula
C7H4BrFO
InChI Key
PJNILWKRAKKEQM-UHFFFAOYSA-N
SMILES
FC1=C(C=O)C(Br)=CC=C1
Assay (HPLC)
>97.5%

Description

It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text