Thermo Scientific Chemicals

9-Bromo-10-phenylanthracene, 98%, Thermo Scientific Chemicals

Catalog number: H64794.03
1 g, Each
Thermo Scientific Chemicals

9-Bromo-10-phenylanthracene, 98%, Thermo Scientific Chemicals

Catalog number: H64794.03
1 g, Each
Quantity
Catalog number: H64794.03
also known as H64794-03
Price (USD)
Price: 67.20
Online price: 57.65
Your price:
Quantity
-

Chemical Identifiers

CAS
23674-20-6
IUPAC Name
9-bromo-10-phenylanthracene
Molecular Formula
C20H13Br
InChI Key
WHGGVVHVBFMGSG-UHFFFAOYSA-N
SMILES
BrC1=C2C=CC=CC2=C(C2=CC=CC=C2)C2=CC=CC=C12
Assay (HPLC)
>97.5%

Description

It is used as an OLED material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an OLED material.

Solubility
Insoluble in water. Soluble in hot toluene, dimethylformamide.

Notes
Store in cool dry place. Keep container tightly closed. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text