1-(2-Thienyl)piperidine, 98%, Thermo Scientific Chemicals
1-(2-Thienyl)piperidine, 98%, Thermo Scientific Chemicals
1-(2-Thienyl)piperidine, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1-(2-Thienyl)piperidine, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H66330.03
also known as H66330-03
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Specifications
Beilstein1239029
CAS19983-20-1
Chemical Name or Material1-(2-Thienyl)piperidine
ColorOrange
Formula Weight167.27
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only