Thermo Scientific Chemicals

Pyruvonitrile, tech. 90%, Thermo Scientific Chemicals

Catalog number: H66636.06
5 g, Each
Thermo Scientific Chemicals

Pyruvonitrile, tech. 90%, Thermo Scientific Chemicals

Catalog number: H66636.06
5 g, Each
Quantity
Catalog number: H66636.06
also known as H66636-06
Price (USD)
Price: 190.00
Online price: 162.65
Your price:
Quantity
-

Chemical Identifiers

CAS
631-57-2
IUPAC Name
acetyl cyanide
Molecular Formula
C3H3NO
InChI Key
QLDHWVVRQCGZLE-UHFFFAOYSA-N
SMILES
CC(=O)C#N
Form
Liquid
Appearance (Color)
Yellow to brown
Assay (GC)
≥88.0%

Description

Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

Solubility
Soluble in ether and acetonitrile.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text