ITIC-F, Thermo Scientific Chemicals
ITIC-F, Thermo Scientific Chemicals
ITIC-F, Thermo Scientific Chemicals
Thermo Scientific Chemicals

ITIC-F, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
250 mg
500 mg
Catalog number H66664.MD
also known as H66664-MD
Price (USD)
916.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,078.00
Save 161.35 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
916.65
Special offer
Online exclusive
Ends: 30-Jun-2026
1,078.00
Save 161.35 (15%)
Each
Chemical Identifiers
CAS2097998-59-7
SpecificationsSpecification SheetSpecification Sheet
FormFlake or threadlike
Bandgap energy1.55 ? 0.02 eV
Appearance (Color)Black
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only