Thermo Scientific Chemicals

Magnesium acetate, 1M aq. soln., Thermo Scientific Chemicals

Catalog number: J60041.AE
100 mL, Each
Thermo Scientific Chemicals

Magnesium acetate, 1M aq. soln., Thermo Scientific Chemicals

Catalog number: J60041.AE
100 mL, Each
Quantity
Catalog number: J60041.AE
also known as J60041-AE
Price (USD)
Price: 84.10
Online price: 71.65
Your price:
Quantity
-

Description

Magnesium acetate is used in molecular biology applications such as batch in vitro transcription of RNA and the crystallization of transcription factor:DNA complexes and proteins via the sitting-drop vapor-diffusion method. Also used as a buffer and to detect sodium and as a catalyst for sulfuric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Magnesium acetate is used in molecular biology applications such as batch in vitro transcription of RNA and the crystallization of transcription factor:DNA complexes and proteins via the sitting-drop vapor-diffusion method. Also used as a buffer and to detect sodium and as a catalyst for sulfuric acid.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text