Thermo Scientific Chemicals

Loperamide hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: J60168.06
5 g, Each
Thermo Scientific Chemicals

Loperamide hydrochloride, 98+%, Thermo Scientific Chemicals

Catalog number: J60168.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J60168.06
also known as J60168-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
34552-83-5
IUPAC Name
hydrogen 4-[4-(4-chlorophenyl)-4-hydroxypiperidin-1-yl]-N,N-dimethyl-2,2-diphenylbutanamide chloride
Molecular Formula
C29H34Cl2N2O2
InChI Key
PGYPOBZJRVSMDS-UHFFFAOYSA-N
SMILES
[H+].[Cl-].CN(C)C(=O)C(CCN1CCC(O)(CC1)C1=CC=C(Cl)C=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder
Loss on Drying
≤0.5%
Appearance (Color)
White to off-white
Assay from Supplier's CofA
≥98.0%
Mass Spec
Consistent with structure

Description

At nanomolar concentrations, binds to mu-opioid receptors. L-type calcium channel blocker

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • At nanomolar concentrations, binds to mu-opioid receptors
  • L-type calcium channel blocker
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text