Thermo Scientific Chemicals

Thymidine-5'-monophosphate disodium salt

Catalog number: J60747.MD
250 mg, Each
Thermo Scientific Chemicals

Thymidine-5'-monophosphate disodium salt

Catalog number: J60747.MD
250 mg, Each
Quantity
Catalog number: J60747.MD
also known as J60747-MD
Price (USD)
Price: 82.70
Online price: 70.65
Your price:
Quantity
-

Chemical Identifiers

CAS
33430-62-5
IUPAC Name
disodium [(2R,3S,5R)-3-hydroxy-5-(5-methyl-2,4-dioxo-1,2,3,4-tetrahydropyrimidin-1-yl)oxolan-2-yl]methyl phosphate
Molecular Formula
C10H13N2Na2O8P
InChI Key
AGSQMPPRYZYDFV-ZJWYQBPBSA-L
SMILES
[Na+].[Na+].CC1=CN([C@H]2C[C@H](O)[C@@H](COP([O-])([O-])=O)O2)C(=O)NC1=O
Form
Powder
Assay (HPLC)
≥ 99.0%
Assay (UV/vis)
≥ 97.0% (Dry basis)
Water Content (Karl Fischer Titration)
≤ 13%
Solution Test
Clear colorless solution (50 mg/ml in water)

Description

Thymidine-5'-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Thymidine-5′-monophosphate disodium salt is a nucleoside monophosphate used to prepare thymidine diphosphate by thymidylate kinase (TMPK) which is a nucleoside monophosphate kinase. In addition to this, it serves as a constituent of deoxyribonucleic acid.

Solubility
Soluble in water.

Notes
Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text