Thermo Scientific Chemicals

Glucagon (1-29) trifluoroacetate salt, human

Catalog number: J60827.EXD
0.5 mg, Each
Thermo Scientific Chemicals

Glucagon (1-29) trifluoroacetate salt, human

Catalog number: J60827.EXD
0.5 mg, Each
Quantity
Catalog number: J60827.EXD
also known as J60827-!D
Price (USD)
Price: 183.00
Your price:
Quantity
-
Appearance (Color)
White
Form
Lyophilized powder
Assay from Suppliers CofA
Peptide content: ≥80%
Assay from Suppliers CofA
Peptide purity: ≥95% (HPLC)
Comment
Solubility: 1mg/ml in water

Description

A peptide hormone that plays a role in maintaining glucose homeostasis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text