Thermo Scientific Chemicals

HEPES-buffered saline, pH 6.5 (5X)

Catalog number: J61239.AK
250 mL, Each
Thermo Scientific Chemicals

HEPES-buffered saline, pH 6.5 (5X)

Catalog number: J61239.AK
250 mL, Each
Quantity
Catalog number: J61239.AK
also known as J61239-AK
Price (USD)
Price: 80.00
Online price: 68.65
Your price:
Quantity
-

Chemical Identifiers

CAS
7365-45-9
IUPAC Name
2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonic acid
Molecular Formula
C8H18N2O4S
InChI Key
JKMHFZQWWAIEOD-UHFFFAOYSA-N
SMILES
OCCN1CCN(CCS(O)(=O)=O)CC1
pH
6.35-6.65
Concentration
HEPES: 125 mM
Concentration
Sodium Chloride: 750 mM

Description

HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
HEPES is typically used in cell culture at concentration between 5mM to 30 mM. It is used in cell culture media, in biopharma buffer formulations (both upstream and downstream), and in diagnostic reagents. HEPES based buffers are used in purification bioprocesses of antibodies, bioactives, proteins and growth factors.

Solubility
Fully miscible in water.

Notes
Contains 125mM HEPES and 750mM sodium chloride, pH 6.5
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text