Thermo Scientific Chemicals

Fluorescein disodium salt, Thermo Scientific Chemicals

Catalog number: J61549.22
100 g, Each
Thermo Scientific Chemicals

Fluorescein disodium salt, Thermo Scientific Chemicals

Catalog number: J61549.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61549.22
Price (USD)
Price: 34.00
Online price: 28.65
Your price:
Quantity
-

Chemical Identifiers

CAS
518-47-8
IUPAC Name
disodium 2-(6-oxido-3-oxo-3H-xanthen-9-yl)benzoate
Molecular Formula
C20H10Na2O5
InChI Key
NJDNXYGOVLYJHP-UHFFFAOYSA-L
SMILES
[Na+].[Na+].[O-]C(=O)C1=CC=CC=C1C1=C2C=CC(=O)C=C2OC2=CC([O-])=CC=C12
Loss on Drying
≤ 8.0% (1 hour at 135?C)
Solution Test
Clear to slightly hazy dark brownish yellow solution with greenish fluorescence (1 mg/ml in water)
Appearance (Color)
Dark brownish-red to brownish-orange, dark orange-red
Form
Powder
Assay (special)
≥90.0% (on dry basis)

Description

Fluorescein disodium salt is used as fluorescent tracer, has been utilized in studies as a drug model, as a penetrating dye . Water soluble fluorescent dye used in permeability and vascular perfusion studies due to its small molecular radius.1 Sodium fluorescein is used as a fluorescent tracer to study blood-brain barrier (BBB) and blood-spinal cord barrier (BSCB) permeability in rodent models. The dye was used as a probe substrate in the study of organic anion transporting polypeptide (OATP) mediated drug transport in hepatocytes.5 Sodium fluorescein was also used to monitor in situ solute transportation into bone through fluorescence recovery after photobleaching (FRAP). Additional reported uses include microscopy, as a gain medium in dye laser use and to detect latent blood stains in vitro. Common derivatives of Fluorescein include fluorescein isothiocyanate (FITC), eosin and Rose Bengal (sc-296278) and Rose Bengal sodium salt

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Fluorescein disodium salt is a yellow-green phthalate indicator dye that is pH-sensitive

• It has a maximum absorbance of 494nm and maximum emission of 521nm

Application

• Fluorescein disodium salt can be used as a fluorescent protein labeler

• Fluorescein is also useful for the staining of cells and organelles

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text