Thermo Scientific Chemicals

Cyclopiazonic acid, 98%, Thermo Scientific Chemicals

Catalog number: J61594.MA
10 mg, Each
Thermo Scientific Chemicals

Cyclopiazonic acid, 98%, Thermo Scientific Chemicals

Catalog number: J61594.MA
10 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J61594.MA
also known as J61594-MA
Price (USD)
Quantity
-

Chemical Identifiers

CAS
18172-33-3
Appearance (Color)
White to off-white
Purity
≥97.5% (HPLC)
Form
Solid

Description

The use of Cyclopiazonic acid on Langendorff-perfused murine hearts displayed anti-arrhythmic effects. Also, administration of cyclopiazonic acid on rat soleus muscle showed a decrease in hormone sensitive lipase activity and triacylglycerol hydrolysis. It is also used as an inhibitor of endoplasmic reticulum Ca2+-ATPase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The use of Cyclopiazonic acid on Langendorff-perfused murine hearts displayed anti-arrhythmic effects. Also, administration of cyclopiazonic acid on rat soleus muscle showed a decrease in hormone sensitive lipase activity and triacylglycerol hydrolysis. It is also used as an inhibitor of endoplasmic reticulum Ca2+-ATPase.

Solubility
Soluble in DMSO (100 mM), ethanol (5 mM), DMF (∼20 mg/ml), and 1:1 DMSO:PBS(pH 7.2) (0.5 mg/ml). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature: -20°C Keep in a dry place. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text