Thermo Scientific Chemicals

7-Methoxycoumarin-4-acetic acid, Thermo Scientific Chemicals

Catalog number: J62475.MD
250 mg, Each
Thermo Scientific Chemicals

7-Methoxycoumarin-4-acetic acid, Thermo Scientific Chemicals

Catalog number: J62475.MD
250 mg, Each
Quantity
Catalog number: J62475.MD
also known as J62475-MD
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62935-72-2
Form
Powder
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥97.0%
Melting Point
190.0-193.0°C

Description

7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Methoxycoumarin-4-acetic acid is a precursor to coumarin-type fluorescent reagents. It has been uses as fluorescent probe in preparation of two novel LysB29 selectively labeled fluorescent derivatives of human insulin, in the preparation of cell-penetrating peptide transportan 10 (tp10) and as fluorescent label for peptides.

Solubility
Soluble in acetone and dimethyl sulfoxide.

Notes
Desiccate at -20°C. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text