Thermo Scientific Chemicals

MOPS, 0.5M buffer soln., pH 7.5

Catalog number: J62839.AK
250 mL, Each
Thermo Scientific Chemicals

MOPS, 0.5M buffer soln., pH 7.5

Catalog number: J62839.AK
250 mL, Each
Quantity
Catalog number: J62839.AK
also known as J62839-AK
Price (USD)
Price: 120.00
Online price: 102.65
Your price:
Quantity
-

Description

MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
MOPS is frequently used as a buffering agent in biology and biochemistry. It used widely for polyacrylamide gel electrophoresis. It is used as a A morpholino propanesulfonic acid buffer.

Solubility
Soluble in water.

Notes
Usage above 20 mM in mammalian cell culture work is not recommended. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from Strong oxidizing agents, Strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text