Thermo Scientific Chemicals

Tween 20 washing buffer, 1.0% in PBS (20X)

Catalog number: J62844.K3
2 L, Each
Thermo Scientific Chemicals

Tween 20 washing buffer, 1.0% in PBS (20X)

Catalog number: J62844.K3
2 L, Each
Quantity
Catalog number: J62844.K3
also known as J62844-K3
Price (USD)
Price: 274.00
Online price: 232.65
Your price:
Quantity
-

Description

It is used as wash buffers for ELISA, Western blot and other immunoassays as well as a blocking buffer for plate-based assays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text