Thermo Scientific Chemicals

Sodium borate, 0.5M buffer soln., pH 8.5

Catalog number: J62902.AP
500 mL, Each
Thermo Scientific Chemicals

Sodium borate, 0.5M buffer soln., pH 8.5

Catalog number: J62902.AP
500 mL, Each
Quantity
Catalog number: J62902.AP
also known as J62902-AP
Price (USD)
Price: 128.00
Online price: 108.65
Your price:
Quantity
-

Description

This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
This product is ideally suited for many biological applications, including biochemical, molecular biological, cell biological, histological, and immunological research and lab procedures. It can be used to dilute substances and in coating procedures. Additives such as Tween 20 and milk powder can be used to add function as washing buffer or blocking buffer.

Solubility
It is soluble in water.

Notes
Store at 25 degrees C. This product is relatively stable under this condition. Best result obtained with use within 6-8 months of purchase.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text