Thermo Scientific Chemicals

Fexofenadine hydrochloride, Thermo Scientific Chemicals

Catalog number: J63262.MB
25 mg, Each
Thermo Scientific Chemicals

Fexofenadine hydrochloride, Thermo Scientific Chemicals

Catalog number: J63262.MB
25 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J63262.MB
also known as J63262-MB
Price (USD)
Price: 125.00
Online price: 108.65
Your price:
Quantity
-

Chemical Identifiers

CAS
153439-40-8
IUPAC Name
hydrogen 2-(4-{1-hydroxy-4-[4-(hydroxydiphenylmethyl)piperidin-1-yl]butyl}phenyl)-2-methylpropanoic acid chloride
Molecular Formula
C32H40ClNO4
InChI Key
RRJFVPUCXDGFJB-UHFFFAOYNA-N
SMILES
[H+].[Cl-].CC(C)(C(O)=O)C1=CC=C(C=C1)C(O)CCCN1CCC(CC1)C(O)(C1=CC=CC=C1)C1=CC=CC=C1
Form
Powder
Mass Spec
Conforms to structure
Appearance (Color)
White to pale cream
Assay from Supplier's CofA
≥98.0% (HPLC)
Proton NMR
Conforms to structure

Description

Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Fexofenadine hydrochloride is used as an antihistamine that inhibits Cox-1, Cox-2, and is a histamine H1 receptor agonist.

Solubility
Soluble in Dimethyl sulfoxide (50 mM), and methanol.

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text