Thermo Scientific Chemicals

Tween 20 washing buffer, 1% in PBS (10X)

Catalog number: J63314.K3
2 L, Each
Thermo Scientific Chemicals

Tween 20 washing buffer, 1% in PBS (10X)

Catalog number: J63314.K3
2 L, Each
Quantity
Catalog number: J63314.K3
also known as J63314-K3
Price (USD)
Price: 184.00
Online price: 156.65
Your price:
Quantity
-
Concentration
Potassium Chloride: 27 mM
Concentration
Potassium Phosphate Monobasic: 17.6 mM
Concentration
Sodium Chloride: 1.37 M
Concentration
Sodium Phosphate DIbasic: 101 mM
Concentration
Tween 20: 1%

Description

Tween-20 is a non-ionic surfactant that is useful for solubilizing membrane proteins during isolation. Tween-20 is also added to buffers for immunohistochemistry. Tween-20 may also help to prevent non-specific antibody binding and function as a blocking agent in immunoblotting assays. It has been used in preextraction of membranes to remove peripheral proteins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text