Thermo Scientific Chemicals

Gossypol-acetic acid, Thermo Scientific Chemicals

Catalog number: J63517.MD
250 mg, Each
Thermo Scientific Chemicals

Gossypol-acetic acid, Thermo Scientific Chemicals

Catalog number: J63517.MD
250 mg, Each
Quantity
Catalog number: J63517.MD
also known as J63517-MD
Price (USD)
Price: 175.00
Online price: 152.65
Your price:
Quantity
-

Chemical Identifiers

CAS
12542-36-8
IUPAC Name
1,1',6,6',7,7'-hexahydroxy-3,3'-dimethyl-5,5'-bis(propan-2-yl)-[2,2'-binaphthalene]-8,8'-dicarbaldehyde; acetic acid
Molecular Formula
C32H34O10
InChI Key
NIOHNDKHQHVLKA-UHFFFAOYSA-N
SMILES
CC(O)=O.CC(C)C1=C(O)C(O)=C(C=O)C2=C(O)C(=C(C)C=C12)C1=C(O)C2=C(C=O)C(O)=C(O)C(C(C)C)=C2C=C1C
Appearance (Color)
Yellow
Form
Powder
Purity
≥95.0% (HPLC)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text