Isradipine, 98+%
Isradipine, 98+%
Isradipine, 98+%
Thermo Scientific Chemicals

Isradipine, 98+%

L-type calcium channel blocker | CAS: 75695-93-1 | C19H21N3O5 | 371.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mg
50 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number J63920.MA
also known as J63920-MA
Price (USD)
116.65
Special offer
Online exclusive
Ends: 30-Jun-2026
137.00
Save 20.35 (15%)
Each
Quantity:
10 mg
Request bulk or custom format
Price (USD)
116.65
Special offer
Online exclusive
Ends: 30-Jun-2026
137.00
Save 20.35 (15%)
Each
Chemical Identifiers
CAS75695-93-1
IUPAC Name3-methyl 5-propan-2-yl 4-(2,1,3-benzoxadiazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Molecular FormulaC19H21N3O5
InChI KeyHMJIYCCIJYRONP-UHFFFAOYNA-N
SMILESCOC(=O)C1=C(C)NC(C)=C(C1C1=CC=CC2=NON=C12)C(=O)OC(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofA≥98.0%
FormPowder or crystals or crystalline powder
Proton NMRConforms
Isradipine is a L-type calcium channel blocker, Isradipine has anti-atherosclerotic effects in animals and improves endothelium-mediated nitric oxide (NO)-dependent vasodilation in vitro. Additional studies indicate that Isradipine may provide a neuroprotective effect in a rat model of focal ischemia and that Isradipine produces a dose-dependent sparing of dopamine neuronal fibers and cell bodies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Isradipine is a L-type calcium channel blocker, Isradipine has anti-atherosclerotic effects in animals and improves endothelium-mediated nitric oxide (NO)-dependent vasodilation in vitro. Additional studies indicate that Isradipine may provide a neuroprotective effect in a rat model of focal ischemia and that Isradipine produces a dose-dependent sparing of dopamine neuronal fibers and cell bodies.

Solubility
Soluble in DMSO

Notes
Protect from heat. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Zuhlke, R.D.; Bouron, A.; Soldatov, N.M.; Reuter, H. Ca2+ channel sensitivity towards the blocker isradipine is affected by alternative splicing of the human α1C subunit gene. FEBS Letters. 1998, 427, (2), 220-224.
  2. Martine Leroueil-Le Verger.; Laurence Fluckiger.; Young-Il Kim.; Maurice Hoffman.; Philippe Maincent. Preparation and characterization of nanoparticles containing an antihypertensive agent. European Journal of Pharmaceutics and Biopharmaceutics. 1998, 46, (2), 137-143.