Thermo Scientific Chemicals
Locostatin, Thermo Scientific Chemicals
Catalog number: J64425.MA
10 mg, Each
Thermo Scientific Chemicals
Locostatin, Thermo Scientific Chemicals
Catalog number: J64425.MA
10 mg, Each
Quantity
Catalog number: J64425.MA
also known as J64425-MA
Price (USD)
Price: 148.00
Online price: 128.65
Your price:
Quantity
-
Description
Locostatin is a cell permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Locostatin is a cell-permeable, potent inhibitor of Raf kinase inhibitor protein (RKIP)/Raf1 kinase interaction and an inhibitor of cell migration