Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, ultrapure, 99.9%

Catalog number: J65594.A7
5 kg, Each
Thermo Scientific Chemicals

Tris(hydroxymethyl)aminomethane, ultrapure, 99.9%

Catalog number: J65594.A7
5 kg, Each
Quantity
Catalog number: J65594.A7
also known as J65594-A7
Price (USD)
Price: 484.00
Online price: 411.65
Your price:
Quantity
-

Chemical Identifiers

CAS
77-86-1
IUPAC Name
2-amino-2-(hydroxymethyl)propane-1,3-diol
Molecular Formula
C4H11NO3
InChI Key
LENZDBCJOHFCAS-UHFFFAOYSA-N
SMILES
NC(CO)(CO)CO
Appearance (Color)
White
Identification (FTIR)
Conforms to structure
pH
10.0 to 11.5 (5%, water)
Assay from Supplier's CofA
≥99.9% (dried basis)
Assay (special)
Iron (Fe): ≤5ppm

Description

Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis. It acts as a precursor for the preparation of polymers, oxazolones and oxazolidines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tris(hydroxymethyl)aminomethane is used as buffers in biochemistry and molecular biology laboratories. It is used as a primary standard to standardize acid solutions for chemical analysis. It finds application in cell membranes to increase its permeability. As an alternative to sodium bicarbonate, it used in the treatment of metabolic acidosis. It acts as a precursor for the preparation of polymers, oxazolones and oxazolidines.

Solubility
Soluble in water, ethylene glycol, methanol, ethanol, dimethyl formamide, acetone, ethyl acetate, olive oil, chloroform and alkaline solution.

Notes
Hygroscopic. Incompatible with bases and strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text