Thermo Scientific Chemicals

3-Azido-1-propylamine, Thermo Scientific Chemicals

Catalog number: J65668.03
1 g, Each
Thermo Scientific Chemicals

3-Azido-1-propylamine, Thermo Scientific Chemicals

Catalog number: J65668.03
1 g, Each
Quantity
Catalog number: J65668.03
also known as J65668-03
Price (USD)
Price: 616.00
Online price: 523.65
Your price:
Quantity
-

Chemical Identifiers

CAS
88192-19-2
IUPAC Name
3-azidopropan-1-amine
Molecular Formula
C3H8N4
InChI Key
OYBOVXXFJYJYPC-UHFFFAOYSA-N
SMILES
NCCCN=[N+]=[N-]
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay from Supplier's CofA
≥95.0% (proton NMR)

Description

Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azido-containing reagent with carbonyl and carboxyl reactivity3-Azido-1-propylamine is used in the preparation of perylenedimides derivative by reacting with 1,6,7,12-tetrachloroperylene-3,4:9,10-tetracarboxyanhydride, which is used in click chemistry.

Solubility
Soluble in DMSO, DMF, DCM, chloroform, THF, and water.Miscible with dimethylsulfoxide, dimethyl formamide, dichloromethane, tetrahydrofuran, chloroform and water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text