Thermo Scientific Chemicals

Leflunomide, Thermo Scientific Chemicals

Catalog number: J65917.ME
500 mg, Each
Thermo Scientific Chemicals

Leflunomide, Thermo Scientific Chemicals

Catalog number: J65917.ME
500 mg, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J65917.ME
also known as J65917-ME
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
75706-12-6
IUPAC Name
5-methyl-N-[4-(trifluoromethyl)phenyl]-1,2-oxazole-4-carboxamide
Molecular Formula
C12H9F3N2O2
InChI Key
VHOGYURTWQBHIL-UHFFFAOYSA-N
SMILES
CC1=C(C=NO1)C(=O)NC1=CC=C(C=C1)C(F)(F)F
Assay (unspecified)
98.0% min.

Description

An immunosuppressive agent. Inhibits T and B cell proliferation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • An immunosuppressive agent
  • Inhibits T and B cell proliferation
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text