Thermo Scientific Chemicals

Maltotriose, 98%

Catalog number: J66491.03
1 g, Each
Thermo Scientific Chemicals

Maltotriose, 98%

Catalog number: J66491.03
1 g, Each
Quantity
Catalog number: J66491.03
also known as J66491-03
Price (USD)
Price: 103.00
Online price: 87.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1109-28-0
IUPAC Name
2-{[4,5-dihydroxy-2-(hydroxymethyl)-6-{[4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C18H32O16
InChI Key
FYGDTMLNYKFZSV-UHFFFAOYNA-N
SMILES
OCC1OC(OC2C(O)C(O)C(OC3C(O)C(O)C(O)OC3CO)OC2CO)C(O)C(O)C1O
Appearance (Color)
White to pale cream
Infrared spectrum
Conforms
Melting Point
140-150°C
Form
Powder
Assay from Supplier's CofA
≥98%

Description

Maltotriose is a matrose regulon inducer in Escherichia coli. Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Maltotriose is a matrose regulon inducer in Escherichia coli. Maltotriose has been used in a study to investigate the influence of fourth generation poly(propyleneimine) dendrimers on blood cells.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text