Thermo Scientific Chemicals

Ethionamide, 98%, Thermo Scientific Chemicals

Catalog number: J66582.06
5 g, Each
Thermo Scientific Chemicals

Ethionamide, 98%, Thermo Scientific Chemicals

Catalog number: J66582.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66582.06
also known as J66582-06
Price (USD)
Price: 152.00
Your price:
Quantity
-

Chemical Identifiers

CAS
536-33-4
IUPAC Name
2-ethylpyridine-4-carbothioamide
Molecular Formula
C8H10N2S
InChI Key
AEOCXXJPGCBFJA-UHFFFAOYSA-N
SMILES
CCC1=CC(=CC=N1)C(N)=S
Appearance (Color)
Yellow
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥97.5%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text