Thermo Scientific Chemicals

Chlorimuron ethyl, 95%, Thermo Scientific Chemicals

Catalog number: J66605.06
5 g, Each
Thermo Scientific Chemicals

Chlorimuron ethyl, 95%, Thermo Scientific Chemicals

Catalog number: J66605.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66605.06
also known as J66605-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
90982-32-4
IUPAC Name
ethyl 2-({[(4-chloro-6-methoxypyrimidin-2-yl)carbamoyl]amino}sulfonyl)benzoate
Molecular Formula
C15H15ClN4O6S
InChI Key
NSWAMPCUPHPTTC-UHFFFAOYSA-N
SMILES
CCOC(=O)C1=CC=CC=C1S(=O)(=O)NC(=O)NC1=NC(Cl)=CC(OC)=N1
Appearance (Color)
White to cream
Form
Powder
Infrared spectrum
Conforms
Purity
≥94.0%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text