Thermo Scientific Chemicals

Cycloheximide, 90%

Catalog number: J66665.06
5 g, Each
Thermo Scientific Chemicals

Cycloheximide, 90%

Catalog number: J66665.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: J66665.06
also known as J66665-06
Price (USD)
Quantity
-

Chemical Identifiers

CAS
66-81-9
IUPAC Name
4-[(2R)-2-[(1S,3S,5S)-3,5-dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidine-2,6-dione
Molecular Formula
C15H23NO4
InChI Key
YPHMISFOHDHNIV-QTEFRXOUNA-N
SMILES
[H][C@]1(C[C@@H](C)C[C@H](C)C1=O)[C@H](O)CC1CC(=O)NC(=O)C1
Assay from Supplier's CofA
≥90.0%
Form
Powder or crystals or crystalline powder
Appearance (Color)
White to pale cream

Description

A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • A frequently used lab reagent that inhibits fungal growth by targeting protein synthesis
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text