Thermo Scientific Chemicals

Agarose, high gelling temperature, Molecular Biology Reagent

Catalog number: J66704.22
100 g, Each
Thermo Scientific Chemicals

Agarose, high gelling temperature, Molecular Biology Reagent

Catalog number: J66704.22
100 g, Each
Quantity
Catalog number: J66704.22
also known as J66704-22
Price (USD)
Price: 336.00
Online price: 284.65
Your price:
Quantity
-

Chemical Identifiers

CAS
9012-36-6
IUPAC Name
(2S,3R,4S,5R,6R)-2-{[(1S,3S,4S,5S,8R)-3-{[(2S,3R,4S,5S,6R)-2-{[(1S,3R,4S,5S,8R)-3,4-dihydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-3,5-dihydroxy-6-(hydroxymethyl)oxan-4-yl]oxy}-4-hydroxy-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular Formula
C24H38O19
InChI Key
MJQHZNBUODTQTK-WKGBVCLCSA-N
SMILES
OC[C@H]1O[C@@H](O[C@@H]2[C@@H]3CO[C@H]2[C@H](O)[C@H](O[C@H]2[C@@H](O)[C@@H](CO)O[C@@H](O[C@@H]4[C@@H]5CO[C@H]4[C@H](O)[C@H](O)O5)[C@@H]2O)O3)[C@H](O)[C@@H](O)[C@H]1O
Comment
Electroendosmosis, Mr: 0.09-0.13
Gel strength
Gel Strength at 1%: ≥800g/cm2
Comment
Gelling temperature: 42.0 ±1.5?C
Water Content (Karl Fischer Titration)
≤10.0%

Description

High gel strength allows gel formation at low concentrations. Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • High gel strength allows gel formation at low concentrations
  • Ideal for antibiotic sensitivity assays, RID, double diffusion and other immunological applications
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text