Thermo Scientific Chemicals

Triethyl phosphite, 98%, Thermo Scientific Chemicals

Catalog number: L00339.AP
500 mL, Each
Thermo Scientific Chemicals

Triethyl phosphite, 98%, Thermo Scientific Chemicals

Catalog number: L00339.AP
500 mL, Each
Quantity

Chemical Identifiers

CAS
122-52-1
IUPAC Name
triethyl phosphite
Molecular Formula
C6H15O3P
InChI Key
BDZBKCUKTQZUTL-UHFFFAOYSA-N
SMILES
CCOP(OCC)OCC
Appearance (Color)
Clear, colourless
Refractive Index
1.4110 - 1.4150 @20?C
Form
liquid
Assay (GC)
> 97.5%

Description

Triethyl phosphite is used as a ligand in organometallic chemistry and as a reagent in organic synthesis. It is used as reference to phosphoric acid standard in 31P NMR spectroscopy. It acts as a reducing agent to prepare 2-phenylindazole from o-nitrobenzylidineaniline as well as reacts with electrophiles to get phosphonates. It forms a stable complex with copper(I) iodide. It finds application in a modified Staudinger reaction for the alkyl bromide to an amine through the azide. Further, it is also used in the preparation of 3-phenyl-2-substituted indoles by reacting with beta-nitro styrenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Triethyl phosphite is used as a ligand in organometallic chemistry and as a reagent in organic synthesis. It is used as reference to phosphoric acid standard in 31P NMR spectroscopy. It acts as a reducing agent to prepare 2-phenylindazole from o-nitrobenzylidineaniline as well as reacts with electrophiles to get phosphonates. It forms a stable complex with copper(I) iodide. It finds application in a modified Staudinger reaction for the alkyl bromide to an amine through the azide. Further, it is also used in the preparation of 3-phenyl-2-substituted indoles by reacting with beta-nitro styrenes.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents and strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text