Thermo Scientific Chemicals

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Catalog number: L00982.06
5 g, Each
Thermo Scientific Chemicals

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole, 99+%, Thermo Scientific Chemicals

Catalog number: L00982.06
5 g, Each
Quantity
Catalog number: L00982.06
Price (USD)
Price: 50.60
Online price: 43.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1750-12-5
IUPAC Name
4-amino-3-hydrazinyl-4,5-dihydro-1H-1,2,4-triazole-5-thione
Molecular Formula
C2H6N6S
InChI Key
RDIMQHBOTMWMJA-UHFFFAOYSA-N
SMILES
NNC1=NNC(=S)N1N
Form
Crystals or powder or crystalline powder
Assay (HPLC)
≥99.0%
Identification (FTIR)
Conforms
Appearance (Color)
White to cream to pale brown to pale gray to gray

Description

4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Amino-3-hydrazino-5-mercapto-1,2,4-triazole is a reagent used in the determination of aldehydes and other reactive chemicals.was used in purpald assay for determination of bacterial polysaccharides containing substituted or unsubstituted glycol in residues such as glycerol, ribitol, arabinitol, furanosyl galactose and sialic acid. It was used as chromogen during the determination of catalase activity in small tissue samples. It was used as reagent in determination of formaldehyde. It was used for derivatization of neutral monosaccharides during HPLC.

Solubility
Soluble in Dimethyl sulfoxide. (DMSO)

Notes
Store at -20°C. Avoid contact with heat. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text