Thermo Scientific Chemicals

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 98+%, Thermo Scientific Chemicals

Catalog number: L01693.14
25 g, Each
Thermo Scientific Chemicals

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione, 98+%, Thermo Scientific Chemicals

Catalog number: L01693.14
25 g, Each
Quantity
Catalog number: L01693.14
also known as L01693-14
Price (USD)
Price: 152.00
Online price: 129.65
Your price:
Quantity
-

Chemical Identifiers

CAS
1522-22-1
IUPAC Name
(3Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one
Molecular Formula
C5H2F6O2
InChI Key
ZUSKFDORJCVUSD-UPHRSURJSA-N
SMILES
OC(=C/C(=O)C(F)(F)F)C(F)(F)F
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to white to yellow
Assay (GC)
≥98.0%

Description

1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,1,5,5,5-Hexafluoro-2,4-pentanedione acts as a chelating ligand. It is used as liquid crystal intermediate.

Solubility
Not miscible in water.

Notes
Store away from oxidizing agents, reducing agents and water. Hygroscopic.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text