1,2-Bis(phenylthio)ethane, 98+%
1,2-Bis(phenylthio)ethane, 98+%
1,2-Bis(phenylthio)ethane, 98+%
Thermo Scientific Chemicals

1,2-Bis(phenylthio)ethane, 98+%

CAS: 622-20-8 | C14H14S2 | 246.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number L02019.18
also known as L02019-18
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Quantity:
50 g
Request bulk or custom format
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Chemical Identifiers
CAS622-20-8
IUPAC Name{[2-(phenylsulfanyl)ethyl]sulfanyl}benzene
Molecular FormulaC14H14S2
InChI KeyMHCVYAFXPIMYRD-UHFFFAOYSA-N
SMILESC(CSC1=CC=CC=C1)SC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to cream to yellow
FormCrystals or powder or crystalline powder or lumps
Assay (GC)≥98.0%
1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,2-Bis(phenylthio)ethane, is used for the synthesis of the bismaleimide resin, APO-BMI, used in syntactic foams.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only