Thermo Scientific Chemicals

Hexaethylene glycol, 96%, Thermo Scientific Chemicals

Catalog number: L02161.14
25 g, Each
Thermo Scientific Chemicals

Hexaethylene glycol, 96%, Thermo Scientific Chemicals

Catalog number: L02161.14
25 g, Each
Quantity
Catalog number: L02161.14
also known as L02161-14
Price (USD)
Price: 237.00
Online price: 201.65
Your price:
Quantity
-

Chemical Identifiers

CAS
2615-15-8
IUPAC Name
3,6,9,12,15-pentaoxaheptadecane-1,17-diol
Molecular Formula
C12H26O7
InChI Key
IIRDTKBZINWQAW-UHFFFAOYSA-N
SMILES
OCCOCCOCCOCCOCCOCCO
Assay (Silylated GC)
≥95.0%
Appearance (Color)
Clear colorless to yellow
Refractive Index
1.4610-1.4670 @ 20?C
Form
Liquid

Description

Hexaethylene glycol exhibits antioxidant, antimicrobial and anti-cancer properties. It also shows potential application as functional hydraulic fluids. It is used as a surfactant building block as well as used in biological sample preparation. Further, it is employed in the synthesis of binaphthol-based macrocyclic ethers using intramolecular oxidative coupling with CuCl(OH).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexaethylene glycol exhibits antioxidant, antimicrobial and anti-cancer properties. It also shows potential application as functional hydraulic fluids. It is used as a surfactant building block as well as used in biological sample preparation. Further, it is employed in the synthesis of binaphthol-based macrocyclic ethers using intramolecular oxidative coupling with CuCl(OH).

Solubility
Miscible with chloroform and methanol. 

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text