Thermo Scientific Chemicals

8-Bromoguanosine hydrate, 97%, Thermo Scientific Chemicals

Catalog number: L02992.03
1 g, Each
Thermo Scientific Chemicals

8-Bromoguanosine hydrate, 97%, Thermo Scientific Chemicals

Catalog number: L02992.03
1 g, Each
Quantity
Catalog number: L02992.03
also known as L02992-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
332359-99-6
IUPAC Name
2-amino-8-bromo-9-[(2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]-6,9-dihydro-1H-purin-6-one
Molecular Formula
C10H12BrN5O5
InChI Key
ASUCSHXLTWZYBA-UMMCILCDSA-N
SMILES
NC1=NC2=C(N=C(Br)N2[C@@H]2O[C@H](CO)[C@@H](O)[C@H]2O)C(=O)N1
Form
Crystals or powder or crystalline powder
Optical Rotation
-29.2 ± 0.1? (C=2 in DMSO:Water 1:1)
Assay (HPLC)
≥96.0%
Appearance (Color)
White to pale cream or pink
Water Content (Karl Fischer Titration)
≤12.98%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text