Thermo Scientific Chemicals

Octanoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: L03526.AE
100 mL, Each
Thermo Scientific Chemicals

Octanoyl chloride, 99%, Thermo Scientific Chemicals

Catalog number: L03526.AE
100 mL, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L03526.AE
also known as L03526-AE
Price (USD)
Price: 31.40
Online price: 26.65
Your price:
Quantity
-

Chemical Identifiers

CAS
111-64-8
IUPAC Name
octanoyl chloride
Molecular Formula
C8H15ClO
InChI Key
REEZZSHJLXOIHL-UHFFFAOYSA-N
SMILES
CCCCCCCC(Cl)=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Identification (FTIR)
Conforms
Refractive Index
1.4335-1.4370 @ 20?C
Assay (GC)
≥98.5%

Description

Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Octanoyl chloride is used to produce adhesives. It is used as an acylating agent for a variety of compounds such as sugars (e.g. Sucrose, aromatic compounds (e.g. Anisole and monoglycerides.

Solubility
Soluble in organic solvents (acetone, chloroform, toluene, THF). Hydrolyzes in water.

Notes
Moisture Sensitive. Incompatible with Bases, Alcohols, Amines, Oxidizing agents. Store away from alcohol and amines.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text