Thermo Scientific Chemicals

4-Bromobenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: L03794.09
10 g, Each
Thermo Scientific Chemicals

4-Bromobenzoyl chloride, 98+%, Thermo Scientific Chemicals

Catalog number: L03794.09
10 g, Each
Quantity
Catalog number: L03794.09
also known as L03794-09
Price (USD)
Quantity
-

Chemical Identifiers

CAS
586-75-4
IUPAC Name
4-bromobenzoyl chloride
Molecular Formula
C7H4BrClO
InChI Key
DENKGPBHLYFNGK-UHFFFAOYSA-N
SMILES
ClC(=O)C1=CC=C(Br)C=C1
Assay (GC)
≥98.0%
Melting Point (clear melt)
36.0-42.0?C
Appearance (Color)
White to cream to brown
Form
Crystals or powder or crystalline powder or lumps or fused solid
Identification (FTIR)
Conforms

Description

4-Bromobenzoyl chloride is used in the synthesis of HIV-1 protease inhibitors. It is also used in the synthesis of oxazoles, cyclohexanone derivatives as analgesic and antimicrobial agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzoyl chloride is used in the synthesis of HIV-1 protease inhibitors. It is also used in the synthesis of oxazoles, cyclohexanone derivatives as analgesic and antimicrobial agents.

Solubility
Soluble in methanol. Reacts with water forming Hcl.

Notes
Moisture Sensitive. Store away from strong bases. Incompatible with moisture and bases. Store at 2 - 8°C.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text