Thermo Scientific Chemicals

5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals

Catalog number: L03952.06
5 g, Each
Thermo Scientific Chemicals

5-Bromovaleronitrile, 98+%, Thermo Scientific Chemicals

Catalog number: L03952.06
5 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: L03952.06
also known as L03952-06
Price (USD)
Price: 64.70
Your price:
Quantity
-

Chemical Identifiers

CAS
5414-21-1
IUPAC Name
5-bromopentanenitrile
Molecular Formula
C5H8BrN
InChI Key
NWWWGAKVHCSAEU-UHFFFAOYSA-N
SMILES
BrCCCCC#N
Appearance (Color)
Clear colorless to white to pale yellow
Form
Liquid or viscous liquid
Assay (GC)
≥98.0%
Refractive Index
1.4765-1.4805 @ 20?C

Description

5-Bromovaleronitrile is used to produce 5-Bromo-pentanoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromovaleronitrile is used to produce 5-Bromo-pentanoic acid.

Solubility
Slightly soluble in water (4.1 g/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text