N-Methyl-N-trimethylsilylacetamide, 98%
N-Methyl-N-trimethylsilylacetamide, 98%
N-Methyl-N-trimethylsilylacetamide, 98%
Thermo Scientific Chemicals

N-Methyl-N-trimethylsilylacetamide, 98%

CAS: 7449-74-3 | C6H15NOSi | 145.277 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
Catalog number L04101.04
also known as L04101-04
Price (USD)
30.65
Online Exclusive
33.90
Save 3.25 (10%)
Each
Quantity:
2 g
Request bulk or custom format
Price (USD)
30.65
Online Exclusive
33.90
Save 3.25 (10%)
Each
Chemical Identifiers
CAS7449-74-3
IUPAC NameN-methyl-N-(trimethylsilyl)acetamide
Molecular FormulaC6H15NOSi
InChI KeyQHUOBLDKFGCVCG-UHFFFAOYSA-N
SMILESCN(C(C)=O)[Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Appearance (Color)Clear colorless to yellow
Assay (GC)≥95.0%
Refractive Index1.4370-1.4410 @ 20°C
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only