1,1,2,2-Tetrachloroethane, 98+%
1,1,2,2-Tetrachloroethane, 98+%
1,1,2,2-Tetrachloroethane, 98+%
Thermo Scientific Chemicals

1,1,2,2-Tetrachloroethane, 98+%

CAS: 79-34-5 | C2H2Cl4 | 167.838 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number L04489.36
also known as L04489-36
Price (USD)
-
Quantity:
500 g
Request bulk or custom format
Chemical Identifiers
CAS79-34-5
IUPAC Name1,1,2,2-tetrachloroethane
Molecular FormulaC2H2Cl4
InChI KeyQPFMBZIOSGYJDE-UHFFFAOYSA-N
SMILESClC(Cl)C(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to pale yellow
FormLiquid
Assay from Suppliers CofA≥98.0%
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1,2,2-Tetrachloroethane is used in the preparation of organic chemicals, as a solvent for fats, oils, waxes, resins, cellulose acetate and in certain types of Friedel-Crafts reactions. It is also used to make paints, varnishes, rust removers and other chlorinated hydrocarbons with two carbon atoms. It is utilized in the determination of theobromine in cacao. It is also used as an immersion fluid in crystallography. Further, it is used as an alcohol denaturant, fumigant, dry cleaning agent, in photographic film. In addition to this, it is used as an intermediate in the industrial production of trichloroethylene, tetrachloroethylene and1,2-dichloroethylene.

Solubility
Miscible with water, methanol, ethanol, chloroform and acetone.

Notes
Incompatible with strong oxidizing agents, sodium, sodium oxides, strong bases, potassium, nitrates, 2,4-dinitrophenyl disulfide.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

General References:

  1. Tan, C.; Xiang, H.; He, Q.; Yang, C. A One-Pot Copper(II)-Catalyzed Tandem Synthesis of 2-Substituted Pyrrolo[1,2-b]pyridazin-4(1H)-ones. Eur. J. Org. Chem. 2015, 2015 (17), 3656-3660.
  2. Chen, W.; Li, Y.; Zhu, D.; Zheng, S.; Chen, W. Dehydrochlorination of activated carbon-bound 1,1,2,2-tetrachloroethane: Implications for carbonaceous material-based soil/sediment remediation. Carbon 2014, 78, 578-588.